Assay Details
Target Gene Details
Entrez Gene ID: | 14089 |
Gene Name: | fibroblast activation protein |
Gene Aliases: |
SIMP |
Location: |
Chr.2:62500936-62575508 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fap | NM_007986.3 | 8 | 735 | NP_032012.1 |
| AK051959.1 | 8 | 655 | ||
| AK136578.1 | 7 | 725 | ||
| BC019190.1 | 8 | 639 | AAH19190.1 | |
| Y10007.1 | 8 | 735 | CAA71116.1 |
More Information
Set Membership: |
|

Genomic Map