Assay Details
Target Gene Details
Entrez Gene ID: | 67877 |
Gene Name: | N(alpha)-acetyltransferase 20, NatB catalytic subunit |
Gene Aliases: |
1500004D14Rik, 2900026I01Rik, AU041458, D2Ertd186e, Nat5 |
Location: |
Chr.2:145903206-145916425 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa20 | NM_001141965.1 | NP_001135437.1 | ||
| NM_026425.3 | NP_080701.1 | |||
| AK005143.1 | BAB23840.1 | |||
| AK009229.1 | BAB26152.1 | |||
| AK153145.1 | ||||
| BC009157.1 | AAH09157.1 | |||
| BC096451.1 | AAH96451.1 | |||
| BC116374.2 | ||||
| CO802916.1 |
More Information
Set Membership: |
|

Genomic Map