Assay Details
Target Gene Details
Entrez Gene ID: | 269378 |
Gene Name: | S-adenosylhomocysteine hydrolase |
Gene Aliases: |
AA987153, AL024110, CuBP, SAHH |
Location: |
Chr.2:155059310-155074497 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahcy | NM_016661.3 | NP_057870.3 | ||
| AK151097.1 | BAE30109.1 | |||
| AK153421.1 | BAE31981.1 | |||
| AK169331.1 | BAE41084.1 | |||
| BC086781.1 | AAH86781.1 | |||
| BC108367.1 | AAI08368.1 | |||
| BQ929555.1 | ||||
| L32836.1 | AAA70378.1 |
More Information
Set Membership: |
|

Genomic Map