Assay Details
Target Gene Details
Entrez Gene ID: | 68275 |
Gene Name: | replication protein A1 |
Gene Aliases: |
5031405K23Rik, 70kDa, AA589576, AW557552, RF-A, RP-A, Rpa |
Location: |
Chr.11:75300259-75348383 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpa1 | NM_001164223.1 | NP_001157695.1 | ||
| NM_026653.2 | NP_080929.1 | |||
| AK035084.1 | ||||
| AK148035.1 | BAE28302.1 | |||
| AK150785.1 | BAE29849.1 | |||
| AK151590.1 | BAE30530.1 | |||
| AK152296.1 | BAE31105.1 | |||
| AK153107.1 | ||||
| AK153144.1 | BAE31755.1 | |||
| AK165316.1 | BAE38134.1 | |||
| AK165944.1 | BAE38476.1 | |||
| AK167598.1 | BAE39655.1 | |||
| AK168400.1 | ||||
| AK169599.1 | BAE41250.1 | |||
| AK177196.1 | ||||
| AK177907.1 | ||||
| AK197958.1 | ||||
| AK203654.1 | ||||
| BC019119.1 | AAH19119.1 | |||
| CJ078143.1 | ||||
| CJ086677.1 |
More Information
Set Membership: |
|

Genomic Map