Assay Details
Target Gene Details
Entrez Gene ID: | 69641 |
Gene Name: | WD repeat domain 20 |
Gene Aliases: |
2310040A13Rik, AI549910, Wdr20a |
Location: |
Chr.12:110737231-110804283 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr20 | NM_027149.2 | 1 | 35 | NP_081425.2 |
| AK009711.1 | 1 | 65 | BAB26456.1 | |
| AK052481.1 | ||||
| AK136417.1 | BAE22969.1 | |||
| BC026761.1 | AAH26761.1 |

Genomic Map