Assay Details
Target Gene Details
Entrez Gene ID: | 26950 |
Gene Name: | visinin-like 1 |
Gene Aliases: |
VILIP, Vnsl1 |
Location: |
Chr.12:11325245-11436649 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsnl1 | NM_012038.4 | 3 | 600 | NP_036168.1 |
| AJ489331.1 | 3 | 564 | CAD33793.1 | |
| AK005091.2 | 2 | 163 | ||
| AK144454.1 | 3 | 530 | BAE25898.1 | |
| AK160224.1 | 3 | 532 | BAE35699.1 | |
| AY101375.1 | 2 | 302 | AAM48292.1 | |
| BC046226.1 | 3 | 505 | AAH46226.1 | |
| D21165.1 | 2 | 302 | BAA04701.1 | |
| EU373446.1 | 2 | 302 |

Genomic Map