Assay Details
Target Gene Details
Entrez Gene ID: | 14561 |
Gene Name: | growth differentiation factor 11 |
Gene Aliases: |
BMP-11, Bmp11 |
Location: |
Chr.10:128884546-128891718 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdf11 | NM_010272.1 | 1 | 432 | NP_034402.1 |
| AF092734.1 | 1 | 177 | AAD05267.1 |
More Information
Set Membership: |
|

Genomic Map