Assay Details
Target Gene Details
Entrez Gene ID: | 20962 |
Gene Name: | synaptonemal complex protein 3 |
Gene Aliases: |
Cor1, Scp3 |
Location: |
Chr.10:88459587-88473236 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sycp3 | NM_011517.2 | NP_035647.2 | ||
| AK131949.1 | BAE20894.1 | |||
| BC132079.1 | ||||
| BC138509.1 | ||||
| Y08485.1 | CAA69719.1 |
More Information
Set Membership: |
|

Genomic Map