Assay Details
Target Gene Details
Entrez Gene ID: | 72735 |
Gene Name: | Egfr long non-coding downstream RNA |
Gene Aliases: |
2810442I21Rik, 4930500E12Rik, Fabl |
Location: |
Chr.11:16934709-16951282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eldr | NR_110421.1 | |||
| NR_110422.1 | ||||
| NR_110423.1 | ||||
| NR_110424.1 | ||||
| NR_110425.1 | ||||
| AK013290.1 | ||||
| AK015657.1 | ||||
| AK141825.1 | 2 | 2375 | ||
| AK163834.1 | 1 | 1025 | ||
| AK165613.1 | ||||
| BI990615.1 |

Genomic Map