Assay Details
Target Gene Details
Entrez Gene ID: | 215814 |
Gene Name: | coiled-coil domain containing 28A |
Gene Aliases: |
1700009P13Rik, AI480677 |
Location: |
Chr.10:18213685-18234991 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc28a | NM_144820.3 | NP_659069.2 | ||
| AK148447.1 | BAE28559.1 | |||
| BC027759.1 |
More Information
Set Membership: |
|

Genomic Map