Assay Details
Target Gene Details
Entrez Gene ID: | 56277 |
Gene Name: | transmembrane protein 45a |
Gene Aliases: |
C630002M10Rik, M32486, p19.5 |
Location: |
Chr.16:56805161-56886458 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem45a | NM_019631.3 | 5 | 1025 | NP_062605.2 |
| AK049837.1 | 5 | 1026 | BAC33947.1 | |
| AK081478.1 | 5 | 1029 | BAC38228.1 | |
| BC064715.1 | 5 | 1028 | AAH64715.1 | |
| M32486.1 | 5 | 725 | AAA37113.1 |

Genomic Map