Assay Details
Target Gene Details
Entrez Gene ID: | 107522 |
Gene Name: | endothelin converting enzyme 2 |
Gene Aliases: |
1810009K13Rik, 6330509A19Rik, 9630025D12Rik, BB127715 |
Location: |
Chr.16:20611601-20646480 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ece2 | NM_025462.2 | 3 | 945 | NP_079738.2 |
| NM_177940.1 | NP_808809.1 | |||
| NM_177941.1 | NP_808810.1 | |||
| AF489569.1 | AAO72356.1 | |||
| AF489570.1 | AAO72357.1 | |||
| AK007407.1 | BAB25019.1 | |||
| AK007791.1 | 3 | 947 | BAB25257.1 | |
| BC115541.2 | 3 | 947 | ||
| BE956299.1 | 1 | 56 |
More Information
Set Membership: |
|

Genomic Map