Assay Details
Target Gene Details
Entrez Gene ID: | 26374 |
Gene Name: | ring finger and WD repeat domain 2 |
Gene Aliases: |
AI316802, C80879, Cop1 |
Location: |
Chr.1:159232174-159347638 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfwd2 | NM_011931.3 | NP_036061.1 | ||
| AF151110.2 | AAD51094.2 | |||
| BC082804.1 | AAH82804.1 | |||
| BC158002.1 |
More Information
Set Membership: |
|

Genomic Map