Assay Details
Target Gene Details
Entrez Gene ID: | 16333 |
Gene Name: | insulin I |
Gene Aliases: |
Ins-1, Ins2-rs1 |
Location: |
Chr.19:52264323-52265476 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ins1 | NM_008386.3 | 1 | 112 | NP_032412.3 |
| AK007345.1 | BAB24974.1 | |||
| AK007482.1 | BAB25058.1 | |||
| AK008358.1 | BAB25628.1 | |||
| AK148541.1 | BAE28611.1 | |||
| BC098468.1 | AAH98468.1 | |||
| BP773571.1 | 2 | 513 |
More Information
Set Membership: |
|

Genomic Map