Assay Details
Target Gene Details
Entrez Gene ID: | 232714 |
Gene Name: | maltase-glucoamylase |
Gene Aliases: |
6030407P20Rik, MGA |
Location: |
Chr.6:40628825-40769123 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgam | NM_001171003.1 | 15 | 1806 | NP_001164474.1 |
| EU937529.1 | 15 | 1806 |
More Information
Set Membership: |
|

Genomic Map