Assay Details
Target Gene Details
Entrez Gene ID: | 13835 |
Gene Name: | Eph receptor A1 |
Gene Aliases: |
5730453L17Rik, AL033318, Eph, Esk |
Location: |
Chr.6:42358487-42373310 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha1 | NM_023580.4 | 18 | 3193 | NP_076069.2 |
| AK028478.1 | 18 | 3194 | BAC25971.1 | |
| BC071215.1 | 18 | 3157 | AAH71215.1 |
Target Gene Details
Entrez Gene ID: | 22793 |
Gene Name: | zyxin |
Gene Aliases: |
9530098H06Rik, R75157 |
Location: |
Chr.6:42349621-42360213 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zyx | NM_001289617.1 | NP_001276546.1 | ||
| NM_001289618.1 | NP_001276547.1 | |||
| NM_001289619.1 | NP_001276548.1 | |||
| NM_011777.3 | NP_035907.1 | |||
| AK031415.1 | BAC27394.1 | |||
| AK035743.1 | BAC29173.1 | |||
| AK076491.1 |
More Information
Set Membership: |
|

Genomic Map