Assay Details
Target Gene Details
Entrez Gene ID: | 17194 |
Gene Name: | mannose-binding lectin (protein A) 1 |
Gene Aliases: |
MBL-A, MBP-A, S-MBP |
Location: |
Chr.14:41151456-41159596 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbl1 | NM_010775.2 | NP_034905.1 | ||
| AK034788.1 | ||||
| BC012245.1 | ||||
| BC021762.1 | AAH21762.1 | |||
| D11441.1 | BAA02006.1 | |||
| S42292.1 | AAB19342.1 |
More Information
Set Membership: |
|

Genomic Map