Assay Details
Target Gene Details
Entrez Gene ID: | 353211 |
Gene Name: | prune homolog 2 (Drosophila) |
Gene Aliases: |
6330414G02Rik, A230083H22Rik, A330102H22Rik, Bmcc1, mKIAA0367 |
Location: |
Chr.19:16955670-17223958 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prune2 | NM_181348.4 | 3 | 386 | NP_851993.3 |
| AK020727.1 | 3 | 387 | BAB32192.1 | |
| AK039241.1 | 3 | 387 | BAC30291.1 | |
| AK165582.1 | BAE38273.1 | |||
| AK165913.1 | 3 | 387 | BAE38457.1 |

Genomic Map