Assay Details
Target Gene Details
Entrez Gene ID: | 12488 |
Gene Name: | CD2-associated protein |
Gene Aliases: |
AL024079, C78928, METS-1, Mets1 |
Location: |
Chr.17:42749098-42876712 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd2ap | NM_009847.3 | NP_033977.3 | ||
| AF077003.1 | AAC36099.1 | |||
| AF149092.1 | AAF73150.1 | |||
| AJ459109.1 | CAD30510.1 | |||
| AK045218.1 | ||||
| AK205774.1 | ||||
| BC019744.1 | AAH19744.1 | |||
| BC138374.1 | ||||
| BC157901.1 |

Genomic Map