Assay Details
Target Gene Details
Entrez Gene ID: | 223776 |
Gene Name: | selenoprotein O |
Gene Aliases: |
1300018J18Rik |
Location: |
Chr.15:89089107-89100340 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Selo | NM_027905.2 | 9 | 2105 | NP_082181.2 |
| AK005048.1 | 9 | 2105 | BAB23774.1 | |
| AK185958.1 | ||||
| AK209897.1 | ||||
| BC048849.1 | 6 | 1128 | AAH48849.1 | |
| BC117817.1 | ||||
| BC141293.1 |
Target Gene Details
Entrez Gene ID: | 328580 |
Gene Name: | tubulin, gamma complex associated protein 6 |
Gene Aliases: |
AI843275, D430023H11, mKIAA1669 |
Location: |
Chr.15:89099097-89123208 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubgcp6 | NM_001163319.1 | 25 | 5970 | NP_001156791.1 |
| AK052293.1 | 2 | 933 | ||
| BC057626.1 | 10 | 2877 | AAH57626.1 |
More Information
Set Membership: |
|

Genomic Map