Assay Details
Target Gene Details
Entrez Gene ID: | 26565 |
Gene Name: | phospholipase A2, group X |
Gene Aliases: |
PLA2GX, mGXsPLA2, sPLA2-X |
Location: |
Chr.16:13715057-13730983 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g10 | NM_001291009.1 | 4 | 1192 | NP_001277938.1 |
| NM_011987.3 | 4 | 1113 | NP_036117.1 | |
| NR_110990.1 | 5 | 1212 | ||
| AF166097.2 | 4 | 715 | AAF04498.2 | |
| AK076921.1 | 4 | 513 | BAC36530.1 | |
| AV075661.1 | ||||
| BC028879.1 | 4 | 652 | AAH28879.1 |
More Information
Set Membership: |
|

Genomic Map