Assay Details
Target Gene Details
Entrez Gene ID: | 110460 |
Gene Name: | acetyl-Coenzyme A acetyltransferase 2 |
Gene Aliases: |
AW742799, Tcp-1x, Tcp1-rs1 |
Location: |
Chr.17:12943042-12960725 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acat2 | NM_009338.3 | 9 | 1861 | NP_033364.2 |
| AK037249.1 | 9 | 1848 | BAC29776.1 | |
| AK151751.1 | 9 | 1849 | ||
| AK166813.1 | 9 | 1872 | BAE39040.1 | |
| AK167234.1 | 9 | 1861 | BAE39358.1 | |
| BC004823.1 | 9 | 1822 | ||
| BC012496.1 | 9 | 1837 | ||
| M35797.1 | 6 | 1458 | AAA76575.1 |
More Information
Set Membership: |
|

Genomic Map