Assay Details
Target Gene Details
Entrez Gene ID: | 320174 |
Gene Name: | RIKEN cDNA A830082K12 gene |
Gene Aliases: |
- |
Location: |
Chr.13:78198017-78236564 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A830082K12Rik | NR_045195.1 | |||
| AK082108.1 |
More Information
Set Membership: |
|

Genomic Map