Assay Details
Target Gene Details
Entrez Gene ID: | 670727 |
Gene Name: | predicted gene 15412 |
Gene Aliases: |
OTTMUSG00000021245 |
Location: |
Chr.7:96339483-96341964 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15412 | NR_046043.1 | 2 | 512 | |
| AK131755.1 | 2 | 513 |
Target Gene Details
Entrez Gene ID: | 23966 |
Gene Name: | teneurin transmembrane protein 4 |
Gene Aliases: |
Doc4, ELM2, Odz4, R75022, Ten-m4, l(7)-3Rn, l7Rn3, mKIAA1302 |
Location: |
Chr.7:96171237-96911093 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tenm4 | NM_011858.3 | NP_035988.2 | ||
| AF059485.1 | AAC31807.1 | |||
| AK031375.1 | BAC27373.1 | |||
| AK147329.1 | BAE27851.1 | |||
| AK147579.1 | BAE28005.1 |
More Information
Set Membership: |
|

Genomic Map