Assay Details
Target Gene Details
Entrez Gene ID: | 53602 |
Gene Name: | hippocalcin-like 1 |
Gene Aliases: |
NVP-3, Nvp3, VILIP3, Vnsl3, Vsnl3 |
Location: |
Chr.12:17690750-17791944 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hpcal1 | NM_016677.4 | 4 | 1025 | NP_057886.1 |
| AF085192.1 | 4 | 1028 | AAC35552.1 | |
| AK002849.1 | 4 | 1006 | ||
| AK179780.1 | 1 | 228 | ||
| AK182612.1 | 1 | 216 | ||
| AK184538.1 | 2 | 362 | ||
| AK185469.1 | 1 | 228 | ||
| BC001997.1 | 4 | 1001 | AAH01997.1 |

Genomic Map