Assay Details
Target Gene Details
Entrez Gene ID: | 52829 |
Gene Name: | leucine rich adaptor protein 1-like |
Gene Aliases: |
1110029A09Rik, AV077978, AV175137, D4Bwg0951e, WI-13707, bM350F23.1 |
Location: |
Chr.4:80910686-80954301 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lurap1l | NM_026821.5 | NP_081097.2 | ||
| AK003950.1 | BAB23088.1 | |||
| BC021501.1 | AAH21501.2 | |||
| BC030404.1 | AAH30404.1 |
More Information
Set Membership: |
|

Genomic Map