Assay Details
Target Gene Details
Entrez Gene ID: | 12773 |
Gene Name: | chemokine (C-C motif) receptor 4 |
Gene Aliases: |
C-C CKR-4, CHEMR1, Cmkbr4, LESTR, Sdf1r |
Location: |
Chr.9:114490316-114505473 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccr4 | NM_009916.2 | NP_034046.2 | ||
| AK042013.1 | ||||
| BC117041.1 | ||||
| BC119171.1 | ||||
| U15208.1 | AAA92582.1 | |||
| X90862.1 | CAA62372.1 |
More Information
Set Membership: |
|

Genomic Map