Assay Details
Target Gene Details
Entrez Gene ID: | 20851 |
Gene Name: | signal transducer and activator of transcription 5B |
Gene Aliases: |
- |
Location: |
Chr.11:100780731-100859004 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat5b | NM_001113563.1 | NP_001107035.1 | ||
| NM_011489.3 | NP_035619.3 | |||
| AK045531.1 | ||||
| AK150098.1 | BAE29305.1 | |||
| AK154014.1 | BAE32317.1 | |||
| AK154664.1 | BAE32752.1 | |||
| AY040231.1 | AAK74074.1 | |||
| AY042906.1 | AAL05590.1 | |||
| AY044901.1 | AAK97791.1 | |||
| AY044902.1 | AAK97792.1 | |||
| AY044903.1 | AAK97793.1 | |||
| BC024319.1 | AAH24319.1 | |||
| U21110.2 | AAC52282.1 | |||
| Z48539.1 | CAA88420.1 |
More Information
Set Membership: |
|

Genomic Map