Assay Details
Target Gene Details
Entrez Gene ID: | 16534 |
Gene Name: | potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4 |
Gene Aliases: |
IK1, IKCA1, KCA4, KCa3.1, SK4, SKCas, mIKCa1 |
Location: |
Chr.7:24369833-24385212 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnn4 | NM_001163510.1 | NP_001156982.1 | ||
| NM_008433.4 | NP_032459.3 | |||
| AB128930.1 | ||||
| AF042487.1 | AAC32051.1 | |||
| AF072884.1 | AAC32829.1 | |||
| AK010943.1 | BAB27283.1 | |||
| AK085752.1 | ||||
| AK155910.1 | BAE33499.1 | |||
| BC010274.1 | AAH10274.1 |
More Information
Set Membership: |
|

Genomic Map