Assay Details
Target Gene Details
Entrez Gene ID: | 66498 |
Gene Name: | DET1 and DDB1 associated 1 |
Gene Aliases: |
1500034J01Rik, 1700095J19Rik, R74921 |
Location: |
Chr.8:71469171-71476097 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dda1 | NM_001294258.1 | NP_001281187.1 | ||
| NM_001294259.1 | NP_001281188.1 | |||
| NM_025600.2 | NP_079876.1 | |||
| NR_122114.1 | ||||
| NR_122115.1 | ||||
| AA036394.1 | ||||
| AK006318.1 | BAB24524.1 | |||
| AK007085.1 | ||||
| AK032966.1 | BAC28103.1 | |||
| AK149117.1 | ||||
| AK149980.1 | BAE29210.1 | |||
| AK160132.1 | ||||
| AK168085.1 | BAE40058.1 | |||
| AK186341.1 | ||||
| BB610194.1 | ||||
| BC034701.1 | AAH34701.1 | |||
| BC093523.1 | AAH93523.1 | |||
| BY297077.1 | ||||
| BY316490.1 |
More Information
Set Membership: |
|

Genomic Map