Assay Details
Target Gene Details
Entrez Gene ID: | 75495 |
Gene Name: | MORN repeat containing 5 |
Gene Aliases: |
1700010A17Rik |
Location: |
Chr.2:36049473-36079709 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Morn5 | NM_029309.2 | NP_083585.1 | ||
| AK005808.1 | BAB24248.1 | |||
| BC036965.1 | AAH36965.1 | |||
| BC048673.1 | AAH48673.1 |
More Information
Set Membership: |
|

Genomic Map