Assay Details
Target Gene Details
Entrez Gene ID: | 79560 |
Gene Name: | ubiquitin-like domain containing CTD phosphatase 1 |
Gene Aliases: |
4930527B16Rik, 8430435I17Rik, BC002236 |
Location: |
Chr.11:44454571-44470548 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ublcp1 | NM_024475.5 | NP_077795.2 | ||
| AK043952.1 | BAC31711.1 | |||
| AK050031.1 | BAC34039.1 | |||
| AK147744.1 | BAE28110.1 | |||
| AK153998.1 | ||||
| BC085111.1 | AAH85111.1 | |||
| BC106093.1 | AAI06094.1 | |||
| CJ061422.1 |
More Information
Set Membership: |
|

Genomic Map