Assay Details
Target Gene Details
Entrez Gene ID: | 68953 |
Gene Name: | charged multivesicular body protein 2A |
Gene Aliases: |
1500016L11Rik |
Location: |
Chr.7:13032006-13034777 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp2a | NM_026885.3 | NP_081161.1 | ||
| AK005267.1 | BAB23919.1 | |||
| BC012230.1 | AAH12230.1 | |||
| BY157363.1 |
More Information
Set Membership: |
|

Genomic Map