Assay Details
Target Gene Details
Entrez Gene ID: | 71934 |
Gene Name: | carbonic anhydrase 13 |
Gene Aliases: |
2310075C21Rik, Ca13 |
Location: |
Chr.3:14641727-14663002 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car13 | NM_024495.5 | NP_078771.1 | ||
| AF231123.1 | AAK16672.1 | |||
| AK010166.1 | BAB26742.1 | |||
| AK150871.1 | BAE29922.1 | |||
| AK150897.1 | BAE29942.1 | |||
| AK151519.1 | BAE30468.1 | |||
| AK151978.1 | BAE30845.1 | |||
| AK152124.1 | ||||
| AK153080.1 | BAE31705.1 | |||
| AK153258.1 | BAE31849.1 | |||
| AK153353.1 | BAE31927.1 | |||
| AK162621.1 | BAE36996.1 | |||
| BC064050.1 | AAH64050.1 |
More Information
Set Membership: |
|

Genomic Map