Assay Details
Target Gene Details
Entrez Gene ID: | 58248 |
Gene Name: | RIKEN cDNA 1700123O20 gene |
Gene Aliases: |
MNCb-2990 |
Location: |
Chr.14:54686171-54690742 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700123O20Rik | NM_021437.2 | NP_067412.1 | ||
| AB041657.1 | BAA95104.1 | |||
| AK007261.1 | BAB24921.1 | |||
| AK167989.1 | BAE39980.1 | |||
| AK170890.1 | BAE42097.1 | |||
| BC011283.1 | AAH11283.1 |
More Information
Set Membership: |
|

Genomic Map