Assay Details
Target Gene Details
Entrez Gene ID: | 75173 |
Gene Name: | testis expressed 38 |
Gene Aliases: |
4930544O15Rik, Theg4 |
Location: |
Chr.4:115779831-115782759 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tex38 | NM_029196.1 | NP_083472.1 | ||
| AK016041.1 | BAB30090.1 | |||
| CF982527.1 |
More Information
Set Membership: |
|

Genomic Map