Assay Details
Target Gene Details
Entrez Gene ID: | 72068 |
Gene Name: | CCR4-NOT transcription complex, subunit 2 |
Gene Aliases: |
2600016M12Rik, 2810470K03Rik, AA537049, AA959607, AW557563, C79650 |
Location: |
Chr.10:116485161-116581535 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot2 | NM_001037846.3 | NP_001032935.2 | ||
| NM_001037847.2 | NP_001032936.2 | |||
| NM_028082.2 | NP_082358.2 | |||
| NR_024327.1 | ||||
| AK011231.1 | BAB27481.1 | |||
| AK078121.1 | BAC37134.1 | |||
| AK149767.1 | BAE29072.1 | |||
| AK167834.1 | BAE39856.1 | |||
| BC043133.1 | AAH43133.1 | |||
| BC063105.1 | AAH63105.1 | |||
| BC065171.1 | AAH65171.1 | |||
| BC090624.1 | AAH90624.1 |
More Information
Set Membership: |
|

Genomic Map