Assay Details
Target Gene Details
Entrez Gene ID: | 319636 |
Gene Name: | fibronectin type III and SPRY domain containing 1-like |
Gene Aliases: |
A230072O16Rik, Ccdc10, Csdufd1, Fsd1nl, OTTMUSG00000007002 |
Location: |
Chr.4:53629929-53707009 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fsd1l | NM_001195284.1 | NP_001182213.1 | ||
| BB626514.1 |
More Information
Set Membership: |
|

Genomic Map