Assay Details
Target Gene Details
Entrez Gene ID: | 106143 |
Gene Name: | CGG triplet repeat binding protein 1 |
Gene Aliases: |
AA960172, AL023996, AW045841 |
Location: |
Chr.16:64851891-64859491 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cggbp1 | NM_178647.2 | NP_848762.1 | ||
| AK049631.1 | BAC33847.1 | |||
| AK052195.1 | BAC34877.1 | |||
| AK083450.1 | BAC38920.1 | |||
| AK086053.1 | ||||
| AK196633.1 | ||||
| AK202964.1 | ||||
| BC043097.1 | AAH43097.1 | |||
| BC057053.1 | AAH57053.1 |
More Information
Set Membership: |
|

Genomic Map