Assay Details
Target Gene Details
Entrez Gene ID: | 60345 |
Gene Name: | nuclear receptor interacting protein 2 |
Gene Aliases: |
AW491344, NIX1 |
Location: |
Chr.6:128397625-128409784 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrip2 | NM_001162858.1 | NP_001156330.1 | ||
| NM_021717.2 | NP_068363.2 | |||
| AJ278170.1 | CAB95737.1 | |||
| AK134689.1 | BAE22242.1 | |||
| AK156526.1 | BAE33744.1 | |||
| BC094230.1 | 2 | 1369 | AAH94230.1 | |
| BC115523.1 | ||||
| BY718516.1 |
More Information
Set Membership: |
|

Genomic Map