Assay Details
Target Gene Details
Entrez Gene ID: | 100036537 |
Gene Name: | predicted gene 11149 |
Gene Aliases: |
ENSMUSG00000060603, ENSMUSG00000079564 |
Location: |
Chr.9:49518288-49568025 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11149 | NR_029465.1 | |||
| AK015955.1 |
Target Gene Details
Entrez Gene ID: | 17967 |
Gene Name: | neural cell adhesion molecule 1 |
Gene Aliases: |
CD56, E-NCAM, NCAM-1, Ncam |
Location: |
Chr.9:49502134-49799427 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncam1 | NM_001081445.1 | NP_001074914.1 | ||
| NM_001113204.1 | NP_001106675.1 | |||
| NM_010875.3 | NP_035005.2 | |||
| AK051197.1 | BAC34554.2 | |||
| AK082621.1 | BAC38551.2 | |||
| BC011310.1 | AAH11310.1 | |||
| X15049.1 | CAA33148.1 | |||
| Y00051.1 | CAA68263.1 |
More Information
Set Membership: |
|

Genomic Map