Assay Details
Target Gene Details
Entrez Gene ID: | 22668 |
Gene Name: | splicing factor 1 |
Gene Aliases: |
BBP, MZFM, WBP4, Zfp162 |
Location: |
Chr.19:6363690-6378038 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf1 | NM_001110791.1 | NP_001104261.1 | ||
| NM_011750.2 | NP_035880.2 | |||
| NR_122118.1 | ||||
| AK133895.1 | ||||
| AK146151.1 | BAE26935.1 | |||
| AK147081.1 | BAE27661.1 | |||
| AK152914.1 | ||||
| AK154705.1 | ||||
| AK157845.1 | BAE34225.1 | |||
| AK214037.1 | ||||
| BC009091.1 | AAH09091.1 | |||
| BC055370.1 | AAH55370.1 | |||
| X85802.1 | CAA59797.1 |
More Information
Set Membership: |
|

Genomic Map