Assay Details
Target Gene Details
Entrez Gene ID: | 26409 |
Gene Name: | mitogen-activated protein kinase kinase kinase 7 |
Gene Aliases: |
B430101B05, C87327, Tak1 |
Location: |
Chr.4:31962296-32023470 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k7 | NM_009316.1 | NP_033342.1 | ||
| NM_172688.3 | NP_766276.1 | |||
| AK046565.1 | ||||
| AK053911.1 | BAC35588.1 | |||
| AK137046.1 | BAE23216.1 | |||
| AK145302.1 | ||||
| AK159277.1 | BAE34955.1 | |||
| AK211827.1 | ||||
| BC006665.1 | AAH06665.1 | |||
| D76446.1 | BAA11184.1 |
More Information
Set Membership: |
|

Genomic Map