Assay Details
Target Gene Details
Entrez Gene ID: | 110460 |
Gene Name: | acetyl-Coenzyme A acetyltransferase 2 |
Gene Aliases: |
AW742799, Tcp-1x, Tcp1-rs1 |
Location: |
Chr.17:12943042-12960725 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acat2 | NM_009338.3 | NP_033364.2 | ||
| AK013275.1 | BAB28763.1 | |||
| AK037249.1 | BAC29776.1 | |||
| AK151751.1 | ||||
| AK166813.1 | BAE39040.1 | |||
| AK167234.1 | BAE39358.1 | |||
| BC004823.1 | ||||
| BC012496.1 |
More Information
Set Membership: |
|

Genomic Map