Assay Details
Target Gene Details
Entrez Gene ID: | 16365 |
Gene Name: | immunoresponsive gene 1 |
Gene Aliases: |
AI323667, CAD |
Location: |
Chr.14:103047012-103056573 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irg1 | NM_008392.1 | NP_032418.1 | ||
| AK036446.1 | BAC29433.1 | |||
| AK089891.1 | ||||
| AK152177.1 | BAE31008.1 | |||
| AK152248.1 | BAE31070.1 | |||
| AK152332.1 | BAE31131.1 | |||
| AK152635.1 | BAE31377.1 | |||
| AK153013.1 | ||||
| AK172001.1 | BAE42768.1 | |||
| AK172298.1 | BAE42932.1 | |||
| L38281.1 | AAA74554.1 |
More Information
Set Membership: |
|

Genomic Map