Assay Details
Target Gene Details
Entrez Gene ID: | 15185 |
Gene Name: | histone deacetylase 6 |
Gene Aliases: |
Hd6, Hdac5, Sfc6, mHDA2 |
Location: |
Chr.X:7930120-7947889 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdac6 | NM_001130416.1 | NP_001123888.1 | ||
| NM_010413.3 | NP_034543.3 | |||
| AF006603.2 | AAD09835.2 | |||
| AK148092.1 | ||||
| AK148144.1 | BAE28372.1 | |||
| AK154099.1 | BAE32376.1 | |||
| AK155545.1 | ||||
| AK155546.1 | ||||
| AK212628.1 | ||||
| BC041105.1 | AAH41105.1 |
More Information
Set Membership: |
|

Genomic Map