Assay Details
Target Gene Details
Entrez Gene ID: | 218793 |
Gene Name: | ubiquitin-conjugating enzyme E2E 2 |
Gene Aliases: |
BC016265 |
Location: |
Chr.14:18573563-18894262 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2e2 | NM_144839.1 | NP_659088.1 | ||
| AK187401.1 | ||||
| BC016265.1 | AAH16265.1 |
More Information
Set Membership: |
|

Genomic Map