Assay Details
Target Gene Details
Entrez Gene ID: | 26876 |
Gene Name: | alcohol dehydrogenase 4 (class II), pi polypeptide |
Gene Aliases: |
Adh2 |
Location: |
Chr.3:138415497-138430892 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adh4 | NM_011996.2 | NP_036126.2 | ||
| AJ245750.1 | CAB57455.1 | |||
| AK132994.1 | BAE21459.1 | |||
| BC100729.1 | AAI00730.1 | |||
| BC100730.1 | AAI00731.1 | |||
| BC100736.1 | AAI00737.1 |
More Information
Set Membership: |
|

Genomic Map