Assay Details
Target Gene Details
Entrez Gene ID: | 80915 |
Gene Name: | dual specificity phosphatase 12 |
Gene Aliases: |
1190004O14Rik, AA027408, AW049275, ESTM36, LMW-DSP4, T-DSP4, VH1, mVH1 |
Location: |
Chr.1:170874187-170885711 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp12 | NM_023173.2 | NP_075662.2 | ||
| AF268196.1 | AAG44739.1 | |||
| AF280810.1 | AAK69508.1 | |||
| AF280811.1 | AAK69509.1 | |||
| AK004488.1 | BAB23328.1 | |||
| AK155026.1 | ||||
| AK181232.1 | ||||
| BC099453.1 | AAH99453.1 |
More Information
Set Membership: |
|

Genomic Map