Assay Details
Target Gene Details
Entrez Gene ID: | 66441 |
Gene Name: | mago-nashi homolog B (Drosophila) |
Gene Aliases: |
2010012C16Rik, Mago, Magoh, Magoh-rs1 |
Location: |
Chr.6:131284388-131293244 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Magohb | NM_025564.2 | NP_079840.2 | ||
| AK006303.1 | BAB24516.2 | |||
| AK007769.1 | BAB25244.1 | |||
| AK008200.1 | BAB25528.1 | |||
| AK013310.1 | BAB28784.1 | |||
| BC027505.1 | AAH27505.1 | |||
| KF051005.1 |
More Information
Set Membership: |
|

Genomic Map